Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sulfate standard solution

Sulfate standard solution, 500 ml (NIST), 100 mg/l as SO42-, 500 mL
Catalog # HACH89149
Supplier:   Hach
Sulfate standard solution
Catalog # HACH89149
Supplier:  Hach
CAS Number:  
Sulfate standard solution
Sulfate standard solution

Sulfate standard solution, 500 ml (NIST), 100 mg/l as SO42-, 500 mL

CAS Number:  
Price
38,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solution for accuracy checks in sulfate determinations.

Specifications

Specifications

  • Concentration:
    100 mg/l as SO<sub>4</sub><sup>2-</sup>
  • Pk:
    500 mL
  • Description:
    Sulfate standard solution, 500 ml (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.