Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
Catalog # HEWLG1375-87324
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWLG1375-87324
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
Price
195,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
  • Seq:
    118