Orders LinkContactUsLinkComponent
Capillaries for LC systems
Flexible capillaries, stainless steel, without fittings, 157 μl, 800 mm, I.Ø 0,5 mm
Catalog # HEWL5065-9926
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWL5065-9926
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Flexible capillaries, stainless steel, without fittings, 157 μl, 800 mm, I.Ø 0,5 mm
Price
113,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Flexible capillaries, stainless steel, without fittings, 157 μl, 800 mm, I.Ø 0,5 mm
  • Seq:
    104