Capillary columns, HP-1
on 5" cage, Capillary, HP-1, 0,33 µm, 0,20 mm, 25 m
HEWL19091Z-102E
: Agilent
Excellent general purpose column – "Industry Standard".
- Non-polar
- Wide range of applications
- Superior performance for low molecular weight alcohols (
- High temperature limit
- Bonded and cross-linked
- Solvent rinsable
100% Dimethylpolysiloxane
Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
- Column Format:Capillary
- Film thickness:0,33 µm
- Ø int.:0,20 mm
- Description:on 5" cage
- Length:25 m
- Seq:02
- Phase:HP-1
- Cat. No.:HEWL19091Z-102E



