Order Entry
Germany
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Glass bottle for solids, 1 g
Catalog # SERA19550.02
Marketsource
Supplier:   Serva
Digitonin
Catalog # SERA19550.02
Supplier:  Serva
Marketsource
CAS Number:  11024-24-1
Digitonin

Glass bottle for solids, 1 g

CAS Number:  11024-24-1
Price
433,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications