Base materials
Peptone from meat (pancreatic digest), Glass bottle for solids, 1 kg
Catalog # SERA48619.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
- Suitable as nutrient for laboratory media and industrial fermentation
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Description:Peptone from meat (pancreatic digest)