Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone from meat (enzymatic digest), Glass bottle for solids, 1 kg
Catalog # SERA48618.02
Marketsource
Supplier:   Serva
Base materials
Catalog # SERA48618.02
Supplier:  Serva
Marketsource
CAS Number:  
Base materials
Peptone from meat (enzymatic digest), Glass bottle for solids, 1 kg
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Description:
    Peptone from meat (enzymatic digest)
Specifications