Buffer, sodium acetate buffer solution pH 7 (3 mol/L)
Sodium acetate buffer solution pH 7 (3 mol/l), Glasflasche, 250 mL
Catalog # SERA39573.01
Marketsource
Supplier: Serva
Buffer, sodium acetate buffer solution pH 7 (3 mol/L)
Catalog # SERA39573.01
Supplier: Serva
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Packung:Glasflasche
- VE:250 mL
- Bezeichnung:Sodium acetate buffer solution pH 7 (3 mol/l)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



