Order Entry
Germany
Orders LinkContactUsLinkComponent
Vancomycin hydrochloride

Vancomycin hydrochloride

Glass bottle for solids, 5 g
Catalog # SERA38125.03
Marketsource
Supplier:   Serva
Vancomycin hydrochloride
Catalog # SERA38125.03
Supplier:  Serva
Marketsource
CAS Number:  1404-93-9
Vancomycin hydrochloride

Glass bottle for solids, 5 g

CAS Number:  1404-93-9
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
  • CAS Number:1404-93-9
  • Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
  • MDL Number:MFCD03613611
  • Molecular Weight:1485.71 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.