Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Aroclor solutions

Aroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule, 1 mL
Catalog # REST32086
Supplier:   Restek
Aroclor solutions
Catalog # REST32086
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule, 1 mL

CAS Number:  
Price
56,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

Certifications: CRM

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule
  • Cat. No.:
    REST32086
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    Aroclor 1254
  • CAS No.:
    11097-69-1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.