Order Entry
Germany
Orders LinkContactUsLinkComponent
Aroclor solutions

Aroclor solutions

Aroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule, 1 mL
Catalog # REST32085
Supplier:   Restek
Aroclor solutions
Catalog # REST32085
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule, 1 mL

CAS Number:  
Price
66,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

Certifications: CRM

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule
  • Cat. No.:
    REST32085
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    Aroclor 1254
  • CAS No.:
    11097-69-1

Specifications