Aroclor solutions
Aroclor 1242, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule, 1 mL
Catalog # REST32082
Supplier: Restek
Jump to:
Product Details & Documents
Aroclor standards may exhibit lot-to-lot variation.
Certifications: CRM
Specifications
Specifications
- Pk:1 mL
- Description:Aroclor 1242, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Cat. No.:REST32082
- Formula:C₁₂H₆Cl₄
- Description:Aroclor 1242
- CAS No.:53469-21-9
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



