Aroclor solutions
Aroclor 1232, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL
Catalog # REST32066
Supplier: Restek
Jump to:
Product Details & Documents
Aroclor standards may exhibit lot-to-lot variation.
: CRM
Specifications
Specifications
- Pk:1 mL
- Description:Aroclor 1232, 200 µg/ml, isooctane, 1 ml/ampoule
- Cat. No.:REST32066
- Formula:—
- Description:Aroclor 1232
- CAS No.:11141-16-5



