Aroclor solutions
Aroclor 1016/1260, 1000 µg/ml, hexane, 1 ml/ampoule, 1 mL
Catalog # REST32039
Supplier: Restek
Jump to:
Product Details & Documents
Aroclor standards may exhibit lot-to-lot variation.
: CRM
Specifications
Specifications
- Pk:1 mL
- Description:Aroclor 1016/1260, 1000 µg/ml, hexane, 1 ml/ampoule
- Cat. No.:REST32039
- Formula:—
- CAS No.:—



