Aroclor solutions
Aroclor 1016, 2000 µg/ml, hexane, 1 ml/ampoule, 1 mL
Catalog # REST32006
Supplier: Restek
Jump to:
Product Details & Documents
Aroclor standards may exhibit lot-to-lot variation.
Certifications: CRM
Specifications
Specifications
- Pk:1 mL
- Description:Aroclor 1016, 2000 µg/ml, hexane, 1 ml/ampoule
- Cat. No.:REST32006
- Formula:C₁₂H₇Cl₃
- Description:Aroclor 1016
- CAS No.:12674-11-2



