Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(DES-TRY1,MET5)-ENKEPHALIN0C=COLD PACK 1 * 5 mg
Catalog # ICNA219043305
Supplier:
MP Biomedicals
Expand image
(DES-TRY1,MET5)-ENKEPHALIN0C=COLD PACK 1 * 5 mg
Catalog # ICNA219043305
Supplier:
MP Biomedicals