Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
S-[N-(3-PHENYLPROPYL)-THIOCARBAMOYL]-L-C 1 * 100 mg
ICNA215999680
:
MP Biomedicals
S-[N-(3-PHENYLPROPYL)-THIOCARBAMOYL]-L-C 1 * 100 mg
ICNA215999680
:
MP Biomedicals