Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NOR-1 1 * 1 mg
Catalog # ICNA215982601
Supplier:
MP Biomedicals
Expand image
NOR-1 1 * 1 mg
Catalog # ICNA215982601
Supplier:
MP Biomedicals