0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-STEAROYL-2-ARACHIDONOYL-sn-GLYCEROL 1 * 5 mg
ICNA215899405
:
MP Biomedicals
1-STEAROYL-2-ARACHIDONOYL-sn-GLYCEROL 1 * 5 mg
ICNA215899405
:
MP Biomedicals