Orders LinkContactUsLinkComponent
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
stopper, solid, 29/32 NS, 24/29 NS
Catalog # SCOT286576906
Supplier:   DWK Life Sciences
Adapter, stopper type, DURAN®
Catalog # SCOT286576906
Supplier:  DWK Life Sciences
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
stopper, solid, 29/32 NS, 24/29 NS
Price
390,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    24/29 NS
  • Type:
    stopper, solid