Nitric acid 3 % (m/V) for analysis denatured in ethanol Content: 1 l 1 * 1 L
KRAF07189.3700
Nitric acid 3 % (m/V) for analysis denatured in ethanol Content: 1 l 1 * 1 L
KRAF07189.3700
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Disinfectant solutions, Bacillol AF
: BODE CHEMIE
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection