ContactUsLinkComponent
Iodine solution according to Wijs 0.1 mol ICl/l - 0.2 N solution (acetic acid)for determination of the iodine number according to DI N 53241-1:1995 Content: 1 l 1 * 1 L
Catalog # KRAF05171.3700
Supplier:  Bernd Kraft
Iodine solution according to Wijs 0.1 mol ICl/l - 0.2 N solution (acetic acid)for determination of the iodine number according to DI N 53241-1:1995 Content: 1 l 1 * 1 L
Catalog # KRAF05171.3700
Supplier:  Bernd Kraft