Buffer solution, colour coded, ready to use
4,0, Citrate, Green, 20 °C, Plastic bottle, 1 L
KRAF03787.3000
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:1 L
- Reference temperature:20 °C
- Colour:Green
- pH value:4,0



