Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solution, colour coded, ready to use

7,0, Phosphate, Green, 20 °C, Plastic bottle, 500 mL
Catalog # KRAF03786.2000
Supplier:   Bernd Kraft
Buffer solution, colour coded, ready to use
Catalog # KRAF03786.2000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution, colour coded, ready to use
Buffer solution, colour coded, ready to use

7,0, Phosphate, Green, 20 °C, Plastic bottle, 500 mL

CAS Number:  
Price
30,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • NIST traceable
  • Colour coded with choice of colours for pH values 4,00 and 7,00
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • Colour:
    Green
  • pH value:
    7,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.