Buffer solutions, ready to use
8,0, Phosphate, Colourless, 20 °C, Plastic bottle, 500 mL
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:500 mL
- Reference temperature:20 °C
- Colour:Colourless
- pH value:8,0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



