ContactUsLinkComponent
Sodium carbonate/hydrogen carbonate standard 1000 mg C/l for determination of anorganic carbon (TIC) according to DIN EN 1484:1997, DEV H3 Content: 1 l 1 * 1 L
  KRAF01285.3000
Sodium carbonate/hydrogen carbonate standard 1000 mg C/l for determination of anorganic carbon (TIC) according to DIN EN 1484:1997, DEV H3 Content: 1 l 1 * 1 L
  KRAF01285.3000