Flasks, round bottom, Quickfit® ground joints
100 ml, 14/23 NS, 108 mm
201-8468
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
: ISO 1773, ISO 4797.
- Socket size:14/23 NS
- Capacity:100 ml
- Height:108 mm
- Seq:12
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



