Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Flasks, round bottom, Quickfit® ground joints

25 ml, 19/26 NS, 80 mm
  201-8498
 :   SciLabware
Flasks, round bottom, Quickfit® ground joints
  201-8498
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints

25 ml, 19/26 NS, 80 mm

 
69,30 €
   
1 * 5 ST
 

PYREX®, borosilicate glass, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With white, matt labelling area.

 : ISO 1773, ISO 4797.

 
  • Socket size:
    19/26 NS
  • Capacity:
    25 ml
  • Height:
    80 mm
  • Seq:
    06