Order Entry
Germany
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Glass bottle for solids, 10 mg
Catalog # 10100.
Supplier:   Biotium
D(-)-Luciferin
Catalog # 10100.
Supplier:  Biotium
CAS Number:  2591-17-5
D(-)-Luciferin

Glass bottle for solids, 10 mg

CAS Number:  2591-17-5
Price
135,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • EINECS:219-981-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.