Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

9,0, Borate, Colourless, 20 °C, Plastic bottle, 1 L
  KRAF03088.3000
Buffer solutions, ready to use
  KRAF03088.3000
 :  
Buffer solutions, ready to use
Buffer solutions, ready to use

9,0, Borate, Colourless, 20 °C, Plastic bottle, 1 L

 :  
 
36,80 €
   
1 * 1 L
 
  • NIST traceable

 

 

 

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    9,0