Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS
Catalog # WITG2.647.500
Supplier:   witeg Labortechnik
Separating funnels, Squibb
Catalog # WITG2.647.500
Supplier:  witeg Labortechnik
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS
Price
97,90 €
Log in for availability
1 * 2 ST
Product Details & Documents
Separating funnels, Squibb, made of borosilicate glass 3.3, with NS stopcock plugs and solid lass key. With PE-stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    500 ml
  • Seq:
    04