ASTM colour <0.5 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0069
Supplier: Lovibond Tintometer
ASTM colour <0.5 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0069
Supplier: Lovibond Tintometer
Frequently Bought Together
Accessories for pH/°C meter, handheld
Supplier: Testo
CHEM013 MEASUREMENT STANDARD
Supplier: MARKET SOURCE PART PROCESS
Buffer solutions
Supplier: REAGECON
Aloin, Reference Standard
Supplier: USP
GlycoWorks Phosphoglycan SPE Reagents HILIC 1 * 1 KIT
Supplier: WATERS
NORLIDOCAINE HYDROCHLORIDE (MONOETHYLGLYCINEXYLIDIDE HYDROCHLORIDE) 1 * 10 mg
Supplier: LGC Standards PROMOCHEM
Sulfabenzamide certified 1 * 0,25 g
Supplier: EHRENSTORFER
VION TEST MIX 1 * 1 KIT
Supplier: WATERS
2-Methyl-3-heptanone
Supplier: EHRENSTORFER
Complete Volatiles Kit - WP, Supelco®
Supplier: Merck
PCB-Mix 1 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
pH Standard Solutions, IUPAC, Certified
Supplier: Hach