Orders LinkContactUsLinkComponent
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
USP colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, USP discs: 4/66 A, 4/66 B, 4/66 C, 4/66 D; 2 × W680/OG/40 mm fused cell, with 40 mm path length, AF335
Catalog # 633-0259
Supplier:   Lovibond Tintometer
Comparator kits, Lovibond® system 2000
Catalog # 633-0259
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
USP colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, USP discs: 4/66 A, 4/66 B, 4/66 C, 4/66 D; 2 × W680/OG/40 mm fused cell, with 40 mm path length, AF335
Price
2 500,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
  • Glassware of the appropriate type and path length
Specifications
  • Description:
    USP colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, USP discs: 4/66 A, 4/66 B, 4/66 C, 4/66 D; 2 × W680/OG/40 mm fused cell, with 40 mm path length
  • Type:
    AF335