Order Entry
Germany
Orders LinkContactUsLinkComponent
Double spatulas, micro, one conical end

Double spatulas, micro, one conical end

160 mm, 7 mm
Catalog # RSGA039.160
Supplier:   RSG
Double spatulas, micro, one conical end
Catalog # RSGA039.160
Supplier:  RSG
Double spatulas, micro, one conical end
Double spatulas, micro, one conical end

160 mm, 7 mm

Wechseln zu:
Price
38,90 €
Anmelden für die Verfügbarkeit
1 * 5 ST
Product Details & Documents

Micro double spatulas with one conical end, 7 mm wide. Made of stainless steel.

Spezifikationen:
  • Length:
    160 mm
  • Blade width:
    7 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.