Order Entry
Germany
Orders LinkContactUsLinkComponent
Support rods, polished

Support rods, polished

12 mm, 1000 mm
Catalog # RSGA241.208
Supplier:   RSG
Support rods, polished
Catalog # RSGA241.208
Supplier:  RSG
Support rods, polished
Support rods, polished

12 mm, 1000 mm

Price
60,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Polished rods made from stainless steel 18/10. Available with or without thread.

Specifications
  • Ø:
    12 mm
  • Length:
    1000 mm
  • Seq:
    12