Order Entry
Germany
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

185 mm, 7 mm
Catalog # 231-0400
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0400
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro

185 mm, 7 mm

Price
56,90 €
Log in for availability
1 * 10 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.

  • Both ends have an angled spoon
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    185 mm
  • Blade width:
    7 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.