Orders LinkContactUsLinkComponent
Screw caps for micro tubes
Screw caps for micro tubes
Screw caps for micro tubes, White
Catalog # SARS65.716.022
Marketsource
Supplier:   SARSTEDT
Screw caps for micro tubes
Catalog # SARS65.716.022
Supplier:  SARSTEDT
Marketsource
Screw caps for micro tubes
Screw caps for micro tubes, White
Price
386,00 €
Log in for availability
1 * 10.000 ST
Product Details & Documents
PP, coloured.
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw cap with O-ring seal.
Specifications
  • Colour:
    White
  • Description:
    Screw caps for micro tubes
  • Seq:
    11