Screw caps for micro tubes
Screw caps for micro tubes, Green
Catalog # SARS65.716.005
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, coloured.
- Excellent chemical resistance
- Excellent temperature resistance
Screw cap with O-ring seal.
Specifications
- Colour:Green
- Description:Screw caps for micro tubes