Orders LinkContactUsLinkComponent
Reference electrodes
Reference electrodes
Reference electrode, low maintenance, Silamid® reference system, B 1981 HD, −5...+100 °C, Glass, Referid®, KPG annular gap, 12×120 mm, Screw plug head
Catalog # SCOI285111915
Supplier:   SI Analytics
Reference electrodes
Catalog # SCOI285111915
Supplier:  SI Analytics
Reference electrodes
Reference electrodes
Reference electrode, low maintenance, Silamid® reference system, B 1981 HD, −5...+100 °C, Glass, Referid®, KPG annular gap, 12×120 mm, Screw plug head
Price
296,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Specifications
  • Connection:
    Screw plug head
  • Ø×L:
    12×120 mm
  • Type:
    B 1981 HD
  • Description:
    Reference electrode, low maintenance, Silamid® reference system
  • Material:
    Glass
  • Diaphr.:
    KPG annular gap
  • Electrolyte:
    Referid®
  • Temp.:
    −5...+100 °C