Orders LinkContactUsLinkComponent
2-(4-Bromophenyl)ethanol ≥98%
Glass bottle for solids, 5 g
  A14143.06
2-(4-Bromophenyl)ethanol ≥98%
  A14143.06
 :  4654-39-1
2-(4-Bromophenyl)ethanol ≥98%
2-(4-Bromophenyl)ethanol ≥98%
Glass bottle for solids, 5 g
 :  4654-39-1
110,00 €
1 * 5 g
Purity: 98% CAS No.: 4654-39-1 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 110°(230°F) Refractive Index: 1.5730 Boiling Point: 106-108°/1mm Beilstein Reference: 2079929 MDL No.: MFCD00002897
 :  4-Bromophenethyl alcohol
  •  :4654-39-1
  •  :C₈H₉BrO
  •  :MFCD00002897
  •  :201.06 g/mol
  •  :106…108 °C (1 mmHg)
  •  :36…38 °C
  •  :1.436
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :225-093-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g