ContactUsLinkComponent
Vacuum filter microfunnel assembly, pk/50, Sterile disposable funnel with petri dish, pad, and 0.45 um white gridded membrane filter , use with 2831600 1 * 50 items
Catalog # HACH2831500
Supplier:  Hach
undefined
Vacuum filter microfunnel assembly, pk/50, Sterile disposable funnel with petri dish, pad, and 0.45 um white gridded membrane filter , use with 2831600 1 * 50 items
Catalog # HACH2831500
Supplier:  Hach