Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 500 mL
Catalog # A0386.0500
Supplier:   PanReac AppliChem
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Catalog # A0386.0500
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 500 mL

CAS Number:  
Price
26,97 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TE buffer solution (TRIS-EDTA buffer) pH 8,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.