Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, PBS (1X, Dulbecco's) powder pack

Buffer, PBS (1X, Dulbecco's) powder pack

PBS buffer (1X, Dulbecco's), 10 l, 1 KIT
Catalog # A0964.9010
Supplier:   PanReac AppliChem
Buffer, PBS (1X, Dulbecco's) powder pack
Catalog # A0964.9010
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, PBS (1X, Dulbecco's) powder pack
Buffer, PBS (1X, Dulbecco's) powder pack

PBS buffer (1X, Dulbecco's), 10 l, 1 KIT

CAS Number:  
Price
31,46 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Powder mixture to prepare PBS buffer (1X) according to Dulbecco.

Composed of di-sodium hydrogen phosphate anhydrous (1150 mg/l), potassium chloride (200 mg/l), potassium dihydrogen phosphate (200 mg/l) and sodium chloride (8000 mg/l).

Documents

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    PBS buffer (1X, Dulbecco's), 10 l

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.