Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sodium azide

Plastic bottle for solids, 500 g
Catalog # APPCA1430.0500
Toxic
Environment
Supplier:   PanReac AppliChem
Sodium azide
Catalog # APPCA1430.0500
Supplier:  PanReac AppliChem
CAS Number:  26628-22-8
Sodium azide

Plastic bottle for solids, 500 g

CAS Number:  26628-22-8
Price
154,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:13,08653
  • Melting Pt:275 °C
  • Density:1.85 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:247-852-1
  • UN:1687
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications