Sodium azide
Plastic bottle for solids, 100 g
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: Sodium triazide
- CAS Number:26628-22-8
- Formula:NaN₃
- MDL Number:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck Index:13,08653
- Melting Pt:275 °C
- Density:1.85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g



