Order Entry
Germany
Orders LinkContactUsLinkComponent
Scoops

Scoops

350 mm, 120 mm, White
Catalog # SCRW4280350
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # SCRW4280350
Supplier:  SCHROEDER WILESCO
Scoops
Scoops

350 mm, 120 mm, White

Price
8,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

These coloured scoops made from PP are suitable for food contact.

Specifications
  • Width:
    120 mm
  • Colour:
    White
  • Length:
    350 mm
  • Seq:
    04