Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSUE RNA ISOLATION KIT (50) 1 * 50 Tests
Catalog # MOBO15000-50
Supplier:
MO BIO
Expand image
TISSUE RNA ISOLATION KIT (50) 1 * 50 Tests
Catalog # MOBO15000-50
Supplier:
MO BIO