Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cy2 AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgG + IgM (H+L) 1 * 1 mg
Catalog # JACK115-226-0440
Supplier:
Jackson Immuno Research
Expand image
Cy2 AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgG + IgM (H+L) 1 * 1 mg
Catalog # JACK115-226-0440
Supplier:
Jackson Immuno Research