Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Fluorescein (FITC) AffiniPure Goat Anti-Human IgM, Fc(5mu) Fragment Specific 1 * 2 mg
Catalog # JACK109-095-043
Supplier:
Jackson Immuno Research
Expand image
Fluorescein (FITC) AffiniPure Goat Anti-Human IgM, Fc(5mu) Fragment Specific 1 * 2 mg
Catalog # JACK109-095-043
Supplier:
Jackson Immuno Research