D(-)-Lactic acid
Glass bottle for solids, 1 g
Catalog # BACE4003254.1000
Marketsource
Supplier: BACHEM BIOCHEMICA
D(-)-Lactic acid
Catalog # BACE4003254.1000
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 10326-41-7
Jump to:
Product Details & Documents
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068313
- Molecular Weight:90.08 g/mol
- Storage Temperature:Freezer
- EINECS:233-713-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



